Learn More
Fisherbrand™ Replacement Strip Tube Rotor
Product Code. 11451967 Shop All Corning Products
Change view
Click to view available options
Capacity:
8 x 0.2 or 16 x 0.2 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
11451967 8 x 0.2 or 16 x 0.2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11451967 Supplier Fisherbrand™ Supplier No. C1300RTS
Please to purchase this item. Need a web account? Register with us today!
For PCR strips and individual 0.2mL tubes. Fisherbrand™ Replacement Strip Tube Rotors are for use with mini centrifuges.
  • Prolongs the life of your centrifuge
  • Manufactured from high quality, durable materials
Specifications
Capacity 8 x 0.2 or 16 x 0.2 mL
Max. Tube Diameter 140 mm
Includes Transparent Lid