Learn More
Pacific Handy Cutter Replacement Blade Disposal Bank
Click to view available options
:
Blade Bank; Color: Yellow
Description
- This key component of our Safety First System keeps blades out the reach of customers, off of the ground, out of trash bags and all other potentially hazardous locations.
- Safely contains over 750 used blades and can be easily mounted to walls, posts, pallet racks and more.
- By pairing one of PHC's Blade Dispensers (SPD-017, QBD-324, or QBD-379) to the front of the Blade Bank, you create a convenient blade change station that will increase the safety and efficiency of your staff.
- By using both items together, your employees will be encouraged to properly dispose of loose blades and easily access new replacement blades near their work stations and throughout your facility, ultimately increasing overall productivity.
- The BH-00205 does not include the wall mount bracket.
Specifications
Specifications
| Capacity (English) | 0.8 kg |
| Color | Yellow |
| Autoclavable | Not Autoclavable |
| Material | Plastic |
| Product Type | Sharps Container |