Learn More
Repirinast-d4, TRC
Product Code. 30422452
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422452 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30422452 Supplier Toronto Research Chemicals Supplier No. TRCR1445620.5MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Repirinast-d4
CAS 1329836-95-4
Molecular Formula C20H17D4NO5
Quantity 0.5 mg
Synonym 5,6-Dihydro-7,8-dimethyl-4,5-dioxo-4H-pyrano[3,2-c]quinoline-2-carboxylic Acid 3-Methylbutyl-d4 Ester, MY 5116-d4, Romet-d4
InChI Formula InChI=1S/C20H21NO5/c1-10(2)7-8-25-20(24)15-9-14(22)16-18(26-15)13-6-5-11(3)12(4)17(13)21-19(16)23/h5-6,9-10H,7-8H2,1-4H3,(H,21,23)/i7D2,8D2
SMILES O=C1NC2=C(C)C(C)=CC=C2C3=C1C(C=C(C(OC([2H])([2H])C([2H])([2H])C(C)C)=O)O3)=O
IUPAC Name 3-methylbutyl-1,1,2,2-d4 7,8-dimethyl-4,5-dioxo-5,6-dihydro-4H-pyrano[3,2-c]quinoline-2-carboxylate
Molecular Weight (g/mol) 359.41
Formula Weight 359.17