missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ Tryptic Soy Agar
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
3kg
500g
Description
Remel Tryptic Soy Agar (TSA Agar) (Dehydrated) is used for the cultivation of a wide variety of nonfastidious microorganisms.
Specifications
Specifications
| Description | Tryptic Soy Agar |
| Form | Powder |
| Product Type | Agar |
| Quantity | 500 g |
For Laboratory Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction