Learn More
Thermo Scientific™ Remel™ Levine EMB Agar, Dehydrated
Product Code. 11952892
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11952892 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11952892 Supplier Thermo Scientific™ Supplier No. R453402
Please to purchase this item. Need a web account? Register with us today!
Remel EMB Agar, Levine (Dehydrated) provides selective isolation and differentiation of gram-negative enteric bacilli from clinical and industrial samples.
Remel EMB Agar, Levine (Dehydrated) provides selective isolation and differentiation of gram-negative enteric bacilli from clinical and industrial samples.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: BAM, COMPF, IDF, USP.
Description EMB Agar, Levine
Form Powder
Product Type Agar
Quantity 500 g