Learn More
Thermo Scientific™ Remel™ Gram Iodine
Product Code. 11983322
Click to view available options
Quantity:
5/Pk.
Each
Unit Size:
250mL
4 L
Pack of 5
Product Code. 11983322
Brand: Thermo Scientific™ R40077
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ Remel Gram Iodine is a mordant used in Gram stain procedures.
Thermo Scientific™ Remel Gram Iodine is a mordant used in Gram stain procedures.
  • For qualitative procedures to differentiate gram-positive from gram-negative microorganisms
  • Selected Gram stains are CE marked
TRUSTED_SUSTAINABILITY
Specifications
Type Gram Iodine
Quantity Each
Certifications/Compliance CE Marked
Volume (Metric) 3.785 L