missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ DNase Test Agar, Dehydrated

Product Code. 11922902
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11922902 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11922902 Supplier Thermo Scientific™ Supplier No. R453252

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Detect DNase activity in Staphylococcus spp., enteric gram-negative bacilli and pseudomonads with DNase Test Agar (Dehydrated).

Detect deoxyribonuclease (DNase) activity in microorganisms with Thermo Scientific™ Remel™ DNase Test Agar (Dehydrated).

In 1957, Jeffries et al. developed a medium to demonstrate DNase activity of microorganisms that possess the extracellular enzyme deoxyribonuclease1. The DNase enzyme catalyzes the cleavage of phosphodiester linkages of DNA leading to its degradation. Organisms producing DNase enzymes thus show clear zones around the colonies. DNase Test Agar is used to identify various bacteria like staphylococci, enteric gram-negative bacilli and pseudomonads2-5.

DNase Test Agar is used to detect the deoxyribonuclease (DNase) activity in microorganisms.

  • Easy-to-interpret: After flooding the plate with hydrochloric acid (1N):
    • Positive test – a clear zone around the growth line and the rest of the plate remains opaque.
  • Recommended for identification of variety of bacteria like staphylococci, enteric gram-negative bacilli and pseudomonads2-5.

DNase Test Agar contains casein and soy peptone that supply nitrogen, amino acids and peptides necessary for bacterial growth. Sodium chloride provides essential electrolytes and maintains osmotic equilibrium.

DNase producing organisms depolymerize DNA into mononucleotides and oligonucleotides when 1N HCl is added to DNase Test Agar after incubation. HCl reacts with polymerized DNA in the medium to provide free nucleic acids and a cloudy precipitate. In areas where DNA is depolymerized, a clear zone is formed around the DNase producing colonies.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description DNase Test Agar
Form Powder
Product Type Agar
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.