missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ Bordet Gengou Agar Base
Click to view available options
Quantity:
500 g
Unit Size:
500g
Description
Remel Bordet Gengou Agar Base is used for the isolation of Bordetella pertussis and Bordetella parapertussis.
Specifications
Specifications
| Description | Bordet Gengou Agar Base |
| Form | Powder |
| Product Type | Agar |
| Quantity | 500 g |
For Laboratory Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction