missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brucella abortus Agglutinating Sera

Product Code. 11993332
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11993332 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11993332 Supplier Thermo Scientific™ Supplier No. R30164801

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use Brucella abortus Agglutinating Sera as control antisera in a standard Widal test.

Use Thermo Scientific™ Remel™ Brucella abortus Agglutinating Sera as control antisera in a standard Widal test for epidemiological and diagnostics purposes. Serological tests are based on the fact that antibodies produced in the serum in response to exposure to bacterial antigens will visibly agglutinate with bacteria carrying homologous antigens. Brucella Agglutinating Sera is used as a control to check for satisfactory performance.

Surface antigens of Brucella have two specificities termed A and M. Most Brucella abortus and Brucella suis strains have a high ratio of A antigens to M antigens whereas the reverse is true for Brucella melitensis1,2. The antisera are prepared using representative smooth strains of Brucella abortus and used as control antisera.

  • Ready to use - Prepared reagents
  • Easy to store - Store at 2-8°C until use
  • Easy to interpret - Visible agglutination in the presence of homologous antigens

Brucella abortus Agglutinating Sera supplies 2mL of rabbit sera containing antibodies to Brucella abortus, preserved with 0.5% phenol and supplied ready to use in a dropper bottle. Brucella antisera are used as controls to check that suspensions have satisfactory quality.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Agglutinating Sera, Brucella abortus
CE Marker Yes
Detectable Analytes Brucella abortus Antibodies
Format Vial
Packaging Type 2 mL/Vial
Quantity 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.