Learn More
Thermo Scientific™ Remel™ Agar
Description
Thermo Scientific™ Remel Agar is a general purpose media which provides a nutritious base for cultivation of numerous fungi or yeast in either a plated medium or broth.
Specifications
Specifications
| Description | Agar |
| Form | Powder |
| Product Type | Agar |
| Quantity | 500 g |