Learn More
Thermo Scientific™ Remel™ Agar
Product Code. 15238987
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15238987 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15238987 Supplier Thermo Scientific™ Supplier No. R451012
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
General-purpose media which provides a nutritious base for cultivation of numerous fungi or yeast in either a plated medium or broth.
Thermo Scientific™ Remel Agar is a general purpose media which provides a nutritious base for cultivation of numerous fungi or yeast in either a plated medium or broth.
TRUSTED_SUSTAINABILITY
Specifications
Description Agar
Form Powder
Product Type Agar
Quantity 500 g