missing translation for 'onlineSavingsMsg'
Learn More

RelA/NFkB p65 Antibody Pack, Novus Biologicals™

Product Code. 18659824 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
4 Vials
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18659824 4 Vials Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18659824 Supplier Novus Biologicals Supplier No. NBP320216

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody Pack

RelA/NFkB p65 antibody specifically detects RelA/NFkB p65 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RelA/NFkB p65
Applications Western Blot
Conjugate Unconjugated
Gene Alias MGC131774, nf kb p65, NFkB p65, NF-kB p65, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65
Immunogen See individual datasheets.
Quantity 4 Vials
Regulatory Status RUO
Gene ID (Entrez) 5970
Target Species Human
Content And Storage Storage is content dependent.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.