missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Reinforced Grid

Use these grids with Memmert ovens. Memmert™ Reinforced Grids are designed to hold up to 60kg.

Brand:  Memmert™ B32190

Product Code. 15281942

  • 597.00 € / Each
Only null left
Estimated Shipment: 09-02-2026
Add to basket
This item is not returnable. View return policy

Description

Description

  • Reinforced stainless steel grid
  • Max. loading 60kg
  • From size 450 with guide bars and fixing screws and only in connection with option K1
Specifications

Specifications

Guide Bars And Fixing Screws
Only in connection with option K1
60 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.