Learn More
Regorafénib N-oxyde (M2), MedChemExpress
Product Code. 16451248
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451248 5 mg 5mg
16438808 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16451248 Supplier Medchem Express Supplier No. HYI0678
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
MedChemExpress Regorafénib N-oxyde M2 is an active metabolite of Regorafenib. Regorafenib is a multi-target inhibitor for VEGFR1/2/3, PDGFRβ, Kit, RET and Raf-1 with IC50s of 13/4.2/46, 22, 7, 1.5 and 2.5 nM, respectively.
Regorafénib N-oxyde M2 is an active metabolite of Regorafenib. Regorafenib is a multi-target inhibitor for VEGFR1/2/3, PDGFRβ, Kit, RET and Raf-1 with IC50s of 13/4.2/46, 22, 7, 1.5 and 2.5 nM, respectively.
Specifications
Chemical Name or Material Regorafénib N-oxyde (M2)
CAS 835621-11-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H15ClF4N4O4
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (100.24 mM; Need ultrasonic)
SMILES FC1=CC(OC2=CC=[N+]([O-])C(C(NC)=O)=C2)=CC=C1NC(NC3=CC(C(F)(F)F)=C(Cl)C=C3)=O
Molecular Weight (g/mol) 498.81
Formula Weight 498.81
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death