Learn More
WTW™ Reference Electrolyte
Product Code. 10569431
Change view
Click to view available options
Color:
Colorless
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10569431 Colorless 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10569431 Supplier WTW™ Supplier No. 109702
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Specifications
Color Colorless
Type Reference Electrolyte
Includes 1 Bottle with 250mL KCl solution 3M/L (AgCl saturated)