Learn More
Refametinib, MedChemExpress
Product Code. 16496128
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16496128
Brand: Medchem Express HY14691
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Refametinib (BAY 869766; RDEA119) is an orally available, potent, non-ATP-competitive, selective, allosteric MEK1/MEK2 inhibitor with IC50s of 19 nM and 47 nM, respectively.
Refametinib (BAY 869766
Specifications
Chemical Name or Material Refametinib
CAS 923032-37-5
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C19H20F3IN2O5S
Quantity 10 mg
Synonym BAY 869766 RDEA119
Solubility Information DMSO : ≥ 31 mg/mL (54.16 mM)
SMILES O=S(C1(C[C@H](O)CO)CC1)(NC(C(NC(C(F)=C2)=CC=C2I)=C3F)=C(OC)C=C3F)=O
Molecular Weight (g/mol) 572.34
Formula Weight 572.34
Percent Purity 98.97%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease