missing translation for 'onlineSavingsMsg'
Learn More

Refametinib, MedChemExpress

Product Code. 16486516
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16486516

Brand: Medchem Express HY14691

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Refametinib (BAY 869766; RDEA119) is an orally available, potent, non-ATP-competitive, selective, allosteric MEK1/MEK2 inhibitor with IC50s of 19 nM and 47 nM, respectively.

Refametinib (BAY 869766

Specifications

Chemical Name or Material Refametinib
CAS 923032-37-5
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C19H20F3IN2O5S
Quantity 100 mg
Synonym BAY 869766 RDEA119
Solubility Information DMSO : ≥ 31 mg/mL (54.16 mM)
SMILES O=S(C1(C[C@H](O)CO)CC1)(NC(C(NC(C(F)=C2)=CC=C2I)=C3F)=C(OC)C=C3F)=O
Molecular Weight (g/mol) 572.34
Formula Weight 572.34
Percent Purity 98.97%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.