missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Refametinib

Product Code. p-7217505
Encompass_Preferred
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 17150212

Brand: STEMCELL Technologies 73374

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MEK/ERK Pathway Inhibitor; Inhibits MEK1 and MEK2

  • Refametinib is an inhibitor of both mitogen-activated protein kinase kinases 1 (MEK1) and 2 (MEK2) with IC50 values of 19 and 47 nM respectively
  • It binds in an allosteric site adjacent to the ATP pocket and is selective for MEK1/2 versus 205 other kinases (Iverson et al.)

Specifications

CAS 923032-37-5
Synonym BAY-86-9766; RDEA119
Quantity 10 mg
Molecular Formula C19H20F3IN2O5S
Purity ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.