Learn More
STEMCELL Technologies Refametinib
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
- Refametinib is an inhibitor of both mitogen-activated protein kinase kinases 1 (MEK1) and 2 (MEK2) with IC50 values of 19 and 47 nM respectively
- It binds in an allosteric site adjacent to the ATP pocket and is selective for MEK1/2 versus 205 other kinases (Iverson et al.)
Specifications
Specifications
| CAS | 923032-37-5 |
| Synonym | BAY-86-9766; RDEA119 |
| Quantity | 10 mg |
| Molecular Formula | C19H20F3IN2O5S |
| Purity | ≥95% |