missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poulten & Graf GmbH Red Vinyl Bulb
Description
- For dispensing bottles
- Made of red vinyl
Specifications
Specifications
| Quantity | 10 |
| Type | Vinyl Bulb for dispensing bottles |
| For Use With (Equipment) | Dispensing bottlees |
| Material | Vinyl |
| Color | Red |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?