missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Verenigde Kunststof Bedrijven™ Polypropylene Closure for Plastic Bottles
Description
- PP Closure
- Color RED
- Food-grade
- Foam insert
Specifications
Specifications
| Color | Red |
| Material | Polypropylene |
| No. per Pack | 1 Each |
| Quantity | 1 Each |
| Thread Size (Metric) | 80 mm |
| Closure Color | Red |
| Product Type | Screw Cap |
| For Use With (Equipment) | For white bottle |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?