missing translation for 'onlineSavingsMsg'
Learn More

Red/Red Solvent Flex Peri-Pump Tubing, 2-Bridge, 1.14 mm (0.045 in.), 12/pack

Product Code. 30311936
Click to view available options
Color:
Red/Red
Material:
Solvent Flex
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30311936

Brand: VHG Labs VHGD180207

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Inner 1.14 mm
Material Solvent Flex
For Use With Peristaltic Pumps
Shape 3
Color Red/Red
Flexibility Flexible
Quantity 12/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.