Learn More
Deutsch Neumann™ Red Natural Rubber Spacer
0.65 € - 1.45 €
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
10209020
|
Deutsch Neumann™
2010042 |
27 mm |
1.15 €
Each |
In Stock
Log in to see stock. |
|||||
10084760
|
Deutsch Neumann™
2010052 |
34 mm |
1.45 €
Each |
In Stock
Log in to see stock. |
|||||
10074750
|
Deutsch Neumann™
2010032 |
24 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10044730
|
Deutsch Neumann™
2010012 |
18 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10582595
|
Deutsch Neumann™
2010022 |
20 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Red | |
Natural Rubber | |
1Each | |
1 Each |