Learn More
Tenak™ Red Cardboard Cryobox
Cryobox made of water-repellent cardboard for long-lasting durability.
Brand:  Tenak™ TE22542
Additional Details : Weight : 3.50000kg
 View more versions of this product
Product Code. 15192105
  • 167.00 € / Pack of 36
Estimated Shipment: 02-09-2024
to see stock.
Add to basket
Description
Description
  • Constructed of fibreboard/cardboard
  • Water-repellent for longer durability
  • Can be used with a large range of grid dividers
Specifications
Specifications
Red
Square
136 x 136 x 100 mm
Cardboard
Cryo Box