missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Zebrafish SOD1 / Superoxide Dismutase Protein

A DNA sequence encoding the Danio rerio (Zebrafish) (Brachydanio rerio) SOD1 / Superoxide Dismutase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6714-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16025320

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_571369.1
SOD1 / Superoxide Dismutase Protein
Research Use Only
sod1
Recombinant Protein
E. coli
MVNKAVCVLK GTGEVTGTVY FNQEGEKKPV KVTGEITGLT PGKHGFHVHA FGDNTNGCIS AGPHFNPHDK THGGPTDSVR HVGDLGNVTA DASGVAKIEI EDAMLTLSGQ HSIIGRTMVI HEKEDDLGKG GNEESLKTGN AGGRLACGVI GITQ
Greater than 86.2% as determined by SDS-PAGE.
30553
10 μg
Not determined.
Full Length
Zebrafish
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions