missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Virus CXCL2/GRO beta/MIP-2/CINC-3 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
Description
CXCL2 Protein is fully biologically active when compared to standard. The specific activity is determined by the inhibitory effect on monocyte migration response to human MIP-1 alpha using a concentration range of 1.0ug-10.0ug/mL of viral MIP-2 will inhibit 25ng/mL of human MIP-1 alpha.
Specifications
Specifications
| Accession Number | Q98157 |
| For Use With (Application) | SDS-PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in 20 mM PB, pH 7.4, 150 mM NaCl. |
| Gene ID (Entrez) | 2920 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 8 kDa |
| Quantity | 10 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 1 EU/μg of CXCL2/GRO beta/MIP-2/CINC-3 AS determined by LAL method. |
| Gene Alias | chemokine (C-X-C motif) ligand 2, CINC-2 A, C-X-C Motif Chemokine 2, GRO2, GRO2 oncogene, GROB, gro-beta, Growth-regulated protein beta, Macrophage Inflammatory Protein, Macrophage Inflammatory Protein 2, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MIP2, MIP-2 A, MIP2A, MIP2-alpha, SCYB2 |
| Gene Symbol | CXCL2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction