missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Viral Zika Virus (H/PF/2013) NS1 Protein

Product Code. 18774929
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18774929

Brand: R&D Systems™ 9450ZK100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

R&D Systems™ Recombinant Viral Zika Virus (H/PF/2013) NS1 Protein CF is a non-structural protein 1 that is a viral glycoprotein.

The Recombinant Viral Zika Virus (H/PF/2013) NS1 Protein is derived from HEK293.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AHZ13508
Characterization Bioactivity not tested
For Use With (Application) Flow Cytometry
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Molecular Weight (g/mol) 41kDa
Quantity 100 μg
Source Human embryonic kidney cell,HEK293-derived viral Zika Virus (H/PF/2013) NS1 protein Val796 & Ser799-Ser1148,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10EU per 1μg of the protein by the LAL method.
Product Type Recombinant Protein
Conjugate Unconjugated
Species Virus
Reconstitution Reconstitute at 250μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.