missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Sheep TNF-alpha Protein

A DNA sequence encoding the Ovis aries (Sheep) TNF-alpha, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6812-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16066020

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001020031.1
TNF-alpha Protein
Research Use Only
TNF
Recombinant Protein
Yeast
LRSSSQASNN KPVAHVVANI SAPGQLRWGD SYANALMANG VELKDNQLVV PTDGLYLIYS QVLFRGHGCP STPLFLTHTI SRIAVSYQTK VNILSAIKSP CHRETLEGAE AKPWYEPIYQ GGVFQLEKGD RLSAEINLPE YLDYAESGQV YFGIIAL
Greater than 90% as determined by SDS-PAGE.
443540
200 μg
Not determined.
Partial (see sequence information for more details).
Sheep
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions