missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Ubiquitin-like protein SMT3 Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Ubiquitin-like protein SMT3, was expressed in the hosts and tags indicated.
309.00 € - 3405.00 €
Specifications
Accession Number | NP_010798.1 |
---|---|
Gene ID (Entrez) | 852122 |
Name | S. cerevisiae Ubiquitin-like protein SMT3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16049300
|
enQuireBio™
QP7546-EC-100UG |
100 μg |
985.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16069300
|
enQuireBio™
QP7546-EC-1MG |
1 mg |
3405.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16059300
|
enQuireBio™
QP7546-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079300
|
enQuireBio™
QP7546-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089300
|
enQuireBio™
QP7546-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099300
|
enQuireBio™
QP7546-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_010798.1 | |
S. cerevisiae Ubiquitin-like protein SMT3 Protein | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
852122 | |
Research Use Only | |
SMT3 | |
Recombinant Protein | |
E. coli | |
SDSEVNQEAK PEVKPEVKPE THINLKVSDG SSEIFFKIKK TTPLRRLMEA FAKRQGKEMD SLRFLYDGIR IQADQTPEDL DMEDNDIIEA HREQIGG | |
Greater than 90% as determined by SDS-PAGE. |