missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Prostatic Acid Phosphatase / ACPP Protein
A DNA sequence encoding the Staphylococcus aureus (strain Mu50 / ATCC 700699) Prostatic Acid Phosphatase / ACPP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6412-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
WP_000426914.1 | |
S. aureus Prostatic Acid Phosphatase / ACPP Protein | |
Research Use Only | |
acpP | |
Recombinant Protein | |
Yeast | |
MENFDKVKDI IVDRLGVDAD KVTEDASFKD DLGADSLDIA ELVMELEDEF GTEIPDEEAE KINTVGDAVK FINSLEK | |
Greater than 90% as determined by SDS-PAGE. |
31213993 | |
50 μg | |
Not determined. | |
Full Length | |
S. aureus | |
His | |
Tris-based buffer, 50% glycerol |