missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus fabD Protein
A DNA sequence encoding the Staphylococcus aureus (strain COL) fabD, was expressed in the hosts and tags indicated.
309.00 € - 3405.00 €
Specifications
Accession Number | WP_000047348.1 |
---|---|
Name | S. aureus fabD Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | fabD |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16070051
|
enQuireBio™
QP7663-EC-100UG |
100 μg |
985.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16090051
|
enQuireBio™
QP7663-EC-1MG |
1 mg |
3405.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16080051
|
enQuireBio™
QP7663-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16000061
|
enQuireBio™
QP7663-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16010061
|
enQuireBio™
QP7663-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16020061
|
enQuireBio™
QP7663-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_000047348.1 | |
Research Use Only | |
fabD | |
Recombinant Protein | |
E. coli | |
MSKTAIIFPG QGAQKVGMAQ DLFNNNDQAT EILTSAANTL DFDILETMFT DEEGKLGETE NTQPALLTHS SALLAALKNL NPDFTMGHSL GEYSSLVAAD VLSFEDAVKI VRKRGQLMAQ AFPTGVGSMA AVLGLDFDKV DEICKSLSSD DKIIEPANIN CPGQIVVSGH KALIDELVEK GKSLGAKRVM PLAVSGPFHS SLMKVIEEDF SSYINQFEWR DAKFPVVQNV NAQGETDKEV IKSNMVKQLY SPVQFINSTE WLIDQGVDHF IEIGPGKVLS GLIKKINRDV KLTSIQTLED VKGWNEND | |
Greater than 90% as determined by SDS-PAGE. |
S. aureus fabD Protein | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |