missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rubella Virus Rubella Capsid Protein

A cDNA sequence encoding the Rubella Capsid was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13372-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15953359

  • 238.00 € / 100µg
Estimated Shipment: 19-09-2024
to see stock.

Description

Description

Specifications

Specifications

Rubella Virus Rubella Capsid Protein
Research Use Only
Recombinant Protein
E. coli
25mM Tris-HCl pH 8.0, 10mM glutation, 0.2% sarcosyl and 50% glycerol.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
ubella Virus
GST
Rubella protein is >95% pure as determined by 10% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions