missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Rat VEGF120 Protein

Product Code. 18646141 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
500 μg
Unit Size:
1000µg
100µg
10µg
500µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18646141 10 μg 10µg
18666420 500 μg 500µg
18684560 100 μg 100µg
18618061 1000 μg 1000µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18646141 Supplier Novus Biologicals™ Supplier No. NBP27633610ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

VEGF120 protein is fully biologically active when compared to standard. Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. The ED50 for this effect is 2-10 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4.
Gene ID (Entrez) 7422
Molecular Weight (g/mol) MolecularWeight-theroretical: 18.5 kDa
Quantity 10 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 0.1 EU/μg of VEGF120 as determined by LAL method.
Gene Symbol VEGFA
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Inhibitor Properties As a result of glycosylation, it migrates to at least two bands with molecular weights ranging from 18.5 kDa in SDS-PAGE under reducing conditions.
Purity or Quality Grade >95%, by SDS-PAGE
Protein VEGF120
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.