missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
VEGF120 protein is fully biologically active when compared to standard. Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. The ED50 for this effect is 2-10 ng/mL.
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 7422 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 18.5 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 0.1 EU/μg of VEGF120 as determined by LAL method. |
| Gene Symbol | VEGFA |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.) |
| Inhibitor Properties | As a result of glycosylation, it migrates to at least two bands with molecular weights ranging from 18.5 kDa in SDS-PAGE under reducing conditions. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?