missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Sortilin Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Sortilin, was expressed in the hosts and tags indicated.
245.00 € - 2645.00 €
Specifications
Accession Number | NP_113955.1 |
---|---|
Gene ID (Entrez) | 83576 |
Name | Sortilin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16055340
|
enQuireBio™
QP6716-YE-100UG |
100 μg |
770.00 €
100µg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16095340
|
enQuireBio™
QP6716-YE-500UG |
500 μg |
2195.00 €
500µg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16065340
|
enQuireBio™
QP6716-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085340
|
enQuireBio™
QP6716-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16005350
|
enQuireBio™
QP6716-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075340
|
enQuireBio™
QP6716-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_113955.1 | |
Sortilin Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
83576 | |
Research Use Only | |
Sort1 | |
Recombinant Protein | |
Yeast | |
CEENDYTTWL AHSTDPGDYK DGCILGYKEQ FLRLRKSSVC QNGRDYVVAK QPSICPCSLE DFLCDFGYFR PENASECVEQ PELKGHELEF CLYGKEEHLT TNGYRKIPGD RCQGGMNPAR EVKDLKKKCT SNFLNPKKQN SKSSS | |
Greater than 90% as determined by SDS-PAGE. |