missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Procalcitonin Protein

A cDNA sequence encoding the Procalcitonin was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13154-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15952669

  • 7875.00 € / 1mg
Estimated Shipment: 19-09-2024
to see stock.

Description

Description

Specifications

Specifications

Procalcitonin Protein
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHASVPLRSTLESS PGMATLSEEE ARLLAALVQN YMQMKVRELE QEEEQEAEGS SLDSPRSKRC GNLSTCMLGT YTQDLNKFHT FPQTSIGVGA PGKKRDMAKD LETNHHPYFG N
Greater than 95.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Rat
His
Procalcitonin was filtered (0.4microm) and lyophilized in 20mM TRIS and 50mM NaCl, pH 8.0.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions