missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat ITGB1 / Integrin beta-1 / CD29 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) ITGB1 / Integrin beta-1 / CD29, was expressed in the hosts and tags indicated.
245.00 € - 3405.00 €
Specifications
Accession Number | NP_058718.2 |
---|---|
Gene ID (Entrez) | 24511 |
Name | ITGB1 / Integrin beta-1 / CD29 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091860
|
enQuireBio™
QP6242-EC-100UG |
100 μg |
770.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16011870
|
enQuireBio™
QP6242-EC-1MG |
1 mg |
3405.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16001870
|
enQuireBio™
QP6242-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021870
|
enQuireBio™
QP6242-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16031870
|
enQuireBio™
QP6242-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041870
|
enQuireBio™
QP6242-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_058718.2 | |
ITGB1 / Integrin beta-1 / CD29 Protein | |
Not determined. | |
Extracellular Domain | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
24511 | |
Research Use Only | |
Itgb1 | |
Recombinant Protein | |
E. coli | |
QTDKNRCLKA NAKSCGECIQ AGPNCGWCTN TTFLQEGMPT SARCDDLEAL KKKGCHPSDI ENPRGSQTIK KNKNVTNRSK GMAEKLRPED ITQIQPQQLL LKLRSGEPQK FTLKFKRAED YPIDLYYLMD LSYSMKDDLE NVKSLGTDLM NEMRRITSDF RIGFGSFVEK TVMPYISTTP AKLRNPCTSE QNCTSPFSYK NVLSLTDRGE FFNELVGQQR ISGNLDSPEG GFDAIMQVAV CGSLIGWRNV TRLLVFSTDA GFHFAGDGKL GGIVLPNDGQ CHLENNVYTM SHYYDYPSIA HLVQKLSENN IQTIFAVTEE FQPVYKELKN LIPKSAVGTL SGNSSNVIQL IIDAYNSLSS EVILENSKLP DGVTINYKSY CKNGVNGTGE NGRKCSNISI GDEVQFEISI TANKCPNKES ENQLKLNPLG FTEEVEVVLQ FICKCNCQSH GIPASPKCHE GNGTFECGAC RCNEGRVGRH CECSTDEVNS EDMDAYCRKE NSSEICSNNG ECVCGQCVCR KRENTNEIYS GKFCECDNFN CDRSNGLICG GNGVCRCRVC ECYPNYTGSA CDCSLDTVPC VATNGQICNG RGICECGACK CTDPKFQGPT CETCQTCLGV CAEHKECVQC RAFNKGEKKD TCAQECSHFN LTKVESREKL PQPVQVDPVT HCKEKDIDDC WFYFTYSVNS KGEAHVHVVE TPDCPTGPD | |
Greater than 90% as determined by SDS-PAGE. |