missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat GSTA1 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) GSTA1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6122-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16080970

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_058709.2
GSTA1 Protein
Research Use Only
Gsta1
Recombinant Protein
E. coli
SGKPVLHYFN ARGRMECIRW LLAAAGVEFD EKFIQSPEDL EKLKKDGNLM FDQVPMVEID GMKLAQTRAI LNYIATKYDL YGKDMKERAL IDMYTEGILD LTEMIMQLVI CPPDQKEAKT ALAKDRTKNR YLPAFEKVLK SHGQDYLVGN RLTRVDIHLL ELLLYVEEFD ASLLTSFPLL KAFKSRISSL PNVKKFLQPG SQRKLPVDAK QIEEARKIFK F
Greater than 90% as determined by SDS-PAGE.
24422
200 μg
Not determined.
Full Length
Rat
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions