missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Cytochrome P450 1B1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Cytochrome P450 1B1, was expressed in the hosts and tags indicated.
245.00 € - 3405.00 €
Specifications
Accession Number | NP_037072.1 |
---|---|
Gene ID (Entrez) | 25426 |
Name | Cytochrome P450 1B1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16070411
|
enQuireBio™
QP7721-EC-100UG |
100 μg |
770.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16090411
|
enQuireBio™
QP7721-EC-1MG |
1 mg |
3405.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16080411
|
enQuireBio™
QP7721-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16000421
|
enQuireBio™
QP7721-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16010421
|
enQuireBio™
QP7721-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16020421
|
enQuireBio™
QP7721-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_037072.1 | |
Cytochrome P450 1B1 Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
25426 | |
Research Use Only | |
Cyp1b1 | |
Recombinant Protein | |
E. coli | |
MATSLSADSP QQLSSLSTQQ TILLLLVSVL AIVHLGQWLL RQWRRKPWSS PPGPFPWPLI GNAASVGRAS HLYFARLARR YGDVFQIRLG SCPVVVLNGE SAIHQALVQQ GGVFADRPPF ASFRVVSGGR SLAFGHYSER WKERRRAAYG TMRAFSTRHP RSRGLLEGHA LGEARELVAV LVRRCAGGAC LDPTQPIIVA VANVMSAVCF GCRYNHDDAE FLELLSHNEE FGRTVGAGSL VDVMPWLQLF PNPVRTIFRE FEQINRNFSN FVLDKFLRHR ESLVPGAAPR DMMDAFILSA EKKATGDPGD SPSGLDLEDV PATITDIFGA SQDTLSTALL WLLILFTRYP DVQARVQAEL DQVVGRDRLP CMSDQPNLPY VMAFLYESMR FTSFLPVTLP HATTANTFVL GYYIPKNTVV FVNQWSVNHD PAKWSNPEDF DPARFLDKDG FINKALASSV MIFSVGKRRC IGEELSKTLL FLFISILAHQ CNFKANQNEP SNMSFSYGLS IKPKSFKIHV SLRESMKLLD SAVEKLQAEE ACQ | |
Greater than 90% as determined by SDS-PAGE. |