missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Rat CXCL10/IP-10/CRG-2 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Description
CXCL10/INP10 Protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human CXCR3 transfected HEK293 cells is in a concentration range of 10-50 ng/mL.
Specifications
Specifications
| Accession Number | P48973 |
| For Use With (Application) | SDS-PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 3627 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 8.7 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 1 EU/μg of CXCL10/IP-10/CRG-2 AS determined by LAL method. |
| Gene Alias | C7, chemokine (C-X-C motif) ligand 10, crg-2, CXCL10, gIP-10, IFI10, INP10, IP-10, IP-10/CXCL10, mob-1, SCYB10 |
| Gene Symbol | CXCL10 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction