missing translation for 'onlineSavingsMsg'
Learn More

Novex™ Recombinant Protein G - Sepharose™ 4B

Product Code. 10031673
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mL
5 mL
25 mL
Unit Size:
1 L
25mL
2mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10031673 2 mL 1 L
10573395 5 mL 5mL
10078792 2 mL 2mL
10405533 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10031673 Supplier Novex™ Supplier No. 1230061211
This item is not returnable. View return policy

Recombinant Protein G - Sepharose™ 4B

Recombinant protein G is produced in a strain of E. coli which contains the gene for protein G from Streptococcus sp. This product is prepared by covalently coupling highly purified rec-Protein G to Sepharose™ 4B. It is supplied as a 50% suspension containing rec-Protein G-Sepharose™ 4B + PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.1% Proclin).

Applications: Immunoglobulin affinity purification and Immunoprecipitation.

Recombinant protein G binds to the Fc region of many types of immunoglobulin G including:

  • all four subclasses of human IgG
  • mouse IgG
  • rabbit IgG
  • guinea pig IgG
  • rat IgG
  • goat IgG
  • sheep IgG
  • cow IgG
  • horse IgG

    Recombinant protein G does not bind to IgG from chicken or cat, or to human IgA, IgM, or serum albumin. It binds weakly to IgG from dog.
  • Specifications

    Product Type Protein G Sepharose
    Product Line Sepharose
    Column Type Affinity
    Stationary Phase Protein G
    Content And Storage This product is supplied as a 50% suspension (2 ml Recombinant Protein G-Sepharose™ 4B + 2 ml PBS) Wet Ice
    Quantity 2 mL
    Form Suspension

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.