missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Protein A/G/L Protein

Product Code. 18435811 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
25 mg
Unit Size:
1mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18435811 1 mg 1mg
18432802 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18435811 Supplier Novus Biologicals™ Supplier No. NBP234985
  • ONLINE EXCLUSIVE PRICE
    191.00 €
    180.60 € / 1mg
    Save 10.40 €
    5% Off
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Recombinant Protein

Protein A/G/L is a genetically engineered protein containing 5 IgG-binding regions of protein A, 2 of protein G and 5 of Protein L. Cell wall binding region, cell membrane binding region and albumin binding region have been removed from the recombinant Protein A/G/L to ensure the maximum specific IgG binding. The recombinant Protein A/G/L is ideal for purification of polyclonal or monoclonal IgG antibodies. Protein A/G/L binds to human, mouse, rat cow, goat, sheep, rabbit, guinea pig, pig, dog and cat IgG.
TRUSTED_SUSTAINABILITY

Specifications

Concentration LYOPH
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from additive free solution.
Molecular Weight (g/mol) 89.2 kDa
Quantity 1 mg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 0.1 EU/ug of Protein A/G/L as determined by LAL method.
Reconstitution Reconstitute with distilled water or saline.
Research Category Biologically Active Proteins
Purity or Quality Grade >95% by SDS-PAGE and HPLC
Protein Protein A/G/L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.