missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig Uricase Protein
A DNA sequence encoding the Sus scrofa (Pig) Uricase, was expressed in the hosts and tags indicated.
309.00 € - 3405.00 €
Specifications
Accession Number | NP_999435.1 |
---|---|
Gene ID (Entrez) | 397510 |
Name | Uricase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16036440
|
enQuireBio™
QP6871-EC-100UG |
100 μg |
985.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16056440
|
enQuireBio™
QP6871-EC-1MG |
1 mg |
3405.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046440
|
enQuireBio™
QP6871-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066440
|
enQuireBio™
QP6871-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076440
|
enQuireBio™
QP6871-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16086440
|
enQuireBio™
QP6871-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_999435.1 | |
Uricase Protein | |
Not determined. | |
Full Length | |
Pig | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
397510 | |
Research Use Only | |
UOX | |
Recombinant Protein | |
E. coli | |
MAHYRNDYKK NDEVEFVRTG YGKDMIKVLH IQRDGKYHSI KEVATSVQLT LSSKKDYLHG DNSDVIPTDT IKNTVNVLAK FKGIKSIETF AVTICEHFLS SFKHVIRAQV YVEEVPWKRF EKNGVKHVHA FIYTPTGTHF CEVEQIRNGP PVIHSGIKDL KVLKTTQSGF EGFIKDQFTT LPEVKDRCFA TQVYCKWRYH QGRDVDFEAT WDTVRSIVLQ KFAGPYDKGE YSPSVQKTLY DIQVLTLGQV PEIEDMEISL PNIHYLNIDM SKMGLINKEE VLLPLDNPYG RITGTVKRKL TSRL | |
Greater than 90% as determined by SDS-PAGE. |