missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig Transcobalamin-1 Protein
A DNA sequence encoding the Sus scrofa (Pig) Transcobalamin-1, was expressed in the hosts and tags indicated.
469.00 € - 1705.00 €
Specifications
Name | Transcobalamin-1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | TCN1 |
Protein Length | Full Length of Mature Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16048271
|
enQuireBio™
QP9423-IV-100UG |
100 μg |
1705.00 €
100µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16068271
|
enQuireBio™
QP9423-IV-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058271
|
enQuireBio™
QP9423-IV-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Transcobalamin-1 Protein | |
Not determined. | |
Full Length of Mature Protein | |
Pig | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
TCN1 | |
Recombinant Protein | |
E. coli | |
CVVSEKDYSH LRLLISAMDN LEQIRGIYGA SILLSQRLAG IQNPSLEEEL SQRIQDDMNR RDMSNLTSGQ LALIILAFGA CKTPDVRFIH DHHLVEKLGE KFKEEIKNME IHNSNPLTNY YQLSFDVLTL CLFRGNYSIS NVTHYFNPEN KNFNLSGHFS VDTGAVAVLA LTCVKRSISN GKIKAAIKDS DTIQKYIESL VHKIQSEKMV VSLETRIAQE KLCRLSLSHQ TITKMNQIAK KLWTRCLTHS QGVFRLPIAA AQILPALLGK TYLDVTKLLL VPKVQVNITD EPVPVVPTLS PENISVIYCV KINEISNCIN ITVFLDVMKA AQEKNSTIYG FTMTETPWGP YITSVQGIWA NNNERTYWEH SEQQQITKPR SMGIMLSKME SI | |
Greater than 80% as determined by SDS-PAGE. |