missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other PhI p 6 Protein
Specifications
Specifications
| Name | PhI p 6 Protein |
| Quantity | 50 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Species | Baculovirus |
| Protein Tag | His |
| Sequence | MVAMFLAVAV VLGLATSPTA EGGKATTEEQ KLIEDVNASF RAAMATTANV PPADKYKTFE AAFTVSSKRN LADAVSKAPQ LVPKLDEVYN AAYNAADHAA PEDKYEAFVL HFSEALRIIA GTPEVHAVKP GA |
| Buffer | PhI p 6 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?