missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mycoplasma Uncharacterized lipoprotein MPN_083 Protein

A DNA sequence encoding the Mycoplasma pneumoniae (strain ATCC 29342 / M129) Uncharacterized lipoprotein MPN_083, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6955-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036880

  • 1750.00 € / 200µg

Estimated Shipment: 23-11-2022
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_109771.1
Mycoplasma Uncharacterized lipoprotein MPN_083 Protein
200 μg
Not determined.
Partial (see sequence information for more details).
Mycoplasma
His-SUMO
Tris-based buffer, 50% glycerol
877109
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MPN_083
Recombinant Protein
E. coli
CSFKDYIPTP SFRKDFSTEN NFVKNKVPGK DDIYSKFYDL TFSLNFVNNQ AQEFGTGWLI DWKGDENKNL SKNKEGQTAS QTRSSSEQTT DQDANLFTAY IATNLHVADG LKNDQDYAPY NKDGWGQPYP YQQKTQSFLL GKYTKPNVQL VKTNYEKPED AVIEQKLKED SLLFIQTSTL PKTAYAAIDP VNFSYNPTRT NGFWTAGKYN VYNGGNSIGN Y
Videos
SDS
Documents
Certificates
Promotions

Promotions