missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Multi Species PKACa2-RIIa2 Protein

Product Code. 15975278
Click to view available options
Quantity:
1 μg
100 μg
3 μg
Unit Size:
100µg
1µg
3µg
This item is not returnable. View return policy

Product Code. 15975278

Brand: enQuireBio™ QP108241ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the PKACa2-RIIa2 was constructed and used to recombinantly synthesize the protein.

Specifications

Name Multi Species PKACa2-RIIa2 Protein
Quantity 1 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity Holoenzyme can be activated by adding the second messenger cAMP (Activation constant about 100nM) releasing two monomeric catalytic subunits.
Product Type Recombinant Protein
Cross Reactivity Multi Species
Species E. coli
Protein Tag Untagged
Buffer PKA holoenzyme type-II alpha is supplied in 50% glycerol.
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.