missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse TPT1 / Tumor Protein

A DNA sequence encoding the Mus musculus (Mouse) TPT1 / Tumor, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6835-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16056170

  • 3405.00 € / 1mg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_033455.1
TPT1 / Tumor Protein
Research Use Only
Tpt1
Recombinant Protein
Yeast
MIIYRDLISH DELFSDIYKI REIADGLCLE VEGKMVSRTE GAIDDSLIGG NASAEGPEGE GTESTVVTGV DIVMNHHLQE TSFTKEAYKK YIKDYMKSLK GKLEEQKPER VKPFMTGAAE QIKHILANFN NYQFFIGENM NPDGMVALLD YREDGVTPFM IFFKDGLEME KC
Greater than 90% as determined by SDS-PAGE.
22070
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions