missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse TIE1 Fc Protein

Product Code. 15994579
Change view
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15994579 2 μg 2µg
15974579 10 μg 10µg
15984579 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15994579 Supplier enQuireBio™ Supplier No. QP137512ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the TIE1 Fc was constructed and used to recombinantly synthesize the protein.

Specifications

Name TIE1 Fc Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Mouse
Species Mouse
Protein Tag Untagged
Sequence MVWWGSSLLL PTLFLASHVG ASVDLTLLAN LRITDPQRFF LTCVSGEAGA GRSSDPPLLL EKDDRIVRTF PPGQPLYLAR NGSHQVTLRG FSKPSDLVGV FSCVGGAGAR RTRVLYVHNS PGAHLFPDKV THTVNKGDTA VLSAHVHKEK QTDVIWKNNG SYFNTLDWQE ADDGRFQLQL QNVQPPSSGI YSATYLEASP LGSAFFRLIV RGCGAGRWGP GCVKDCPGCL HGGVCHDHDG ECVCPPGFTG TRCEQACREG RFGQSCQEQC PGTAGCRGLT FCLPDPYGCS CGSGWRGSQC QEACAPGHFG ADCRLQCQCQ NGGTCDRFSG CVCPSGWHGV HCEKSDRIPQ ILSMATEVEF NIGTMPRINC AAAGNPFPVR GSMKLRKPDG TMLLSTKVIV EPDRTTAEFE VPSLTLGDSG FWECRVSTSG GQDSRRFKVN VKVPPVPLTA PRLLAKQSRQ LVVSPLVSFS GDGPISSVRL HYRPQDSTIA WSAIVVDPSE NVTLMNLKPK TGYNVRVQLS RPGEGGEGGW GPSALMTTDC PEPLLQPWLE SWHVEGPDRL RVSWSLPSVP LSGDGFLLRL WDGARGQERR ENISFPQART ALLTGLTPGT HYQLDVRLYH CTLLGPASPP AHVHLPPSGP PAPRHLHAQA LSDSEIQLMW QHPEAPSGPI SKYIVEIQVA GGSGDPQWMD VDRPEETSII VRGLNASTRY LFRVRASVQG LGDWSNTVEE ATLGNGLQSE GPVRESRAAE EGLDQQLVLA VVGSVSATCL TILAALLALV CIRRSCLHRR RTFTYQSGSG EETILQFSSG TLTLTRRPKP QPEPLSYPVL EWEDITFEDL IGEGNFGQVI RAMIKKDGLK MNAAIKMLKE YASENDHRDF AGELEVLCKL GHHPNIINLL GACENRGYLY IAIEYAPYGN LLDFLRKSRV LETDPAFARE HGTASTLSSR QLLRFASDAA NGMQYLSEKQ FIHRDLAARN VLVGENLASK IADFGLSRGE EVYVKKTMGR LPVRWMAIES LNYSVYTTKS DVWSFGVLLW EIVSLGGTPY CGMTCAELYE KLPQGYRMEQ PRNCDDEVYE LMRQCWRDRP YERPPFAQIA LQLGRMLEAR KAYVNMSLFE NFTYAGIDAT AEEA
Buffer TIE-1 Fc Chimera was lyophilized from a concentrated (1 mg/ml) sterile solution containing 1x PBS.
Purity or Quality Grade Greater than 90.0% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.