missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Serpina3n Protein
A DNA sequence encoding the Mus musculus (Mouse) Serpina3n, was expressed in the hosts and tags indicated.
313.00 € - 2645.00 €
Specifications
Name | Serpina3n Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | Serpina3n |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091231
|
enQuireBio™
QP7862-EC-10UG |
10 μg |
313.00 €
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16011241
|
enQuireBio™
QP7862-EC-200UG |
200 μg |
1275.00 €
200µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16081231
|
enQuireBio™
QP7862-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16031241
|
enQuireBio™
QP7862-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021241
|
enQuireBio™
QP7862-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001241
|
enQuireBio™
QP7862-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Spécification
Serpina3n Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
Serpina3n | |
Recombinant Protein | |
E. coli | |
FPDGTLGMDA AVQEDHDNGT QLDSLTLASI NTDFAFSLYK ELVLKNPDKN IVFSPLSISA ALAVMSLGAK GNTLEEILEG LKFNLTETSE ADIHQGFGHL LQRLNQPKDQ VQISTGSALF IEKRQQILTE FQEKAKTLYQ AEAFTADFQQ PRQAKKLIND YVRKQTQGMI KELVSDLDKR TLMVLVNYIY FKAKWKVPFD PLDTFKSEFY AGKRRPVIVP MMSMEDLTTP YFRDEELSCT VVELKYTGNA SALFILPDQG RMQQVEASLQ PETLRKWKNS LKPRMIDELH LPKFSISTDY SLEDVLSKLG IREVFSTQAD LSAITGTKDL RVSQVVHKAV LDVAETGTEA AAATGVKFVP MSAKLYPLTV YFNRPFLIMI FDTETEIAPF IAKIANPK | |
Greater than 90% as determined by SDS-PAGE. |