missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Selenoprotein 1

A DNA sequence encoding the Mus musculus (Mouse) Selenoprotein T, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7250-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16097850

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001035486.2
Selenoprotein 1
Research Use Only
Selt
Recombinant Protein
E. coli
SANLGGVPSK RLKMQYATGP LLKFQICVSU GYRRVFEEYM RVISQRYPDI RIEGENYLPQ PIYRHIASFL SVFKLVLIGL IIVGKDPFAF FGMQAPSIWQ WGQENKVYAC MMVFFLSNMI ENQCMSTGAF EITLNDVPVW SKLESGHLPS MQQLVQILDN EMKLNVHMDS IPHHRS
Greater than 90% as determined by SDS-PAGE.
69227
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions